Product Details

SNP ID
rs9486499
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:107030089 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTCTGCTGAATTCCTAGTGCCCA[G/T]ATTAGTGCTTGGCACATGGTAAGTG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C6orf203 PubMed Links

Gene Details

Gene
C6orf203
Gene Name
chromosome 6 open reading frame 203
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142468.2 186 Intron NP_001135940.1
NM_001142470.2 186 UTR 5 NP_001135942.1
NM_016487.4 186 Intron NP_057571.1
XM_011535876.2 186 UTR 5 XP_011534178.1
XM_011535877.2 186 UTR 5 XP_011534179.1
XM_017010919.1 186 Intron XP_016866408.1
XM_017010920.1 186 Intron XP_016866409.1

View Full Product Details