Product Details
- SNP ID
-
rs2114950
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:77430377 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTCATAGCTGGGGAGAAGAAGTTCA[A/G]TGCCAGCGAGTGTAGTTGGTGCCTT
- Phenotype
-
MIM: 603390
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PDE8B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78039653] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PDE8B
- Gene Name
- phosphodiesterase 8B
There are no transcripts associated with this gene.
- Gene
- WDR41
- Gene Name
- WD repeat domain 41
View Full Product Details