Product Details

SNP ID
rs2747098
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:57995513 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATAAACATTTGCTCATTTAATTTT[A/G]GTCAAGTTAAATCAAATGTAAAAAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C14orf37 PubMed Links

Gene Details

Gene
C14orf37
Gene Name
chromosome 14 open reading frame 37
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001872.3 11626 Intron NP_001001872.2
NM_001320173.1 11626 Intron NP_001307102.1
XM_011536471.2 11626 Intron XP_011534773.1
XM_017021029.1 11626 UTR 3 XP_016876518.1
XM_017021030.1 11626 UTR 3 XP_016876519.1

View Full Product Details