Product Details

SNP ID
rs7544381
Assay Type
Functionally tested
NCBI dbSNP Submissions
50
Location
Chr.1:67308610 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCCTTTGTGAACTATGAGGCCCT[C/T]CTCATCTCCCTAGGACACACTAAAT
Phenotype
MIM: 601642
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IL12RB2 PubMed Links

Gene Details

Gene
IL12RB2
Gene Name
interleukin 12 receptor subunit beta 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258214.1 Intron NP_001245143.1
NM_001258215.1 Intron NP_001245144.1
NM_001258216.1 Intron NP_001245145.1
NM_001319233.1 Intron NP_001306162.1
NM_001559.2 Intron NP_001550.1
XM_005270825.2 Intron XP_005270882.1
XM_005270827.2 Intron XP_005270884.1
XM_005270828.3 Intron XP_005270885.1
XM_006710617.2 Intron XP_006710680.1
XM_011541383.2 Intron XP_011539685.1
XM_011541384.2 Intron XP_011539686.1
XM_017001202.1 Intron XP_016856691.1
XM_017001203.1 Intron XP_016856692.1
XM_017001204.1 Intron XP_016856693.1

View Full Product Details