Product Details

SNP ID
rs10819785
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.9:100481700 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GAAGAGCCTGGCGGTACGATGAAAG[A/G]TAATGGTAGAAATGGGAAGAATTGA
Phenotype
MIM: 603421
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
MSANTD3-TMEFF1 PubMed Links

Gene Details

Gene
MSANTD3-TMEFF1
Gene Name
MSANTD3-TMEFF1 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198812.1 Intron NP_001185741.1
Gene
TMEFF1
Gene Name
transmembrane protein with EGF like and two follistatin like domains 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003692.4 Intron NP_003683.2

View Full Product Details