Product Details

SNP ID
rs10989104
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:100476287 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTAGTTGGTAAGTTTGAGAACCAT[A/G]TTAATACAGTCTGGACACACACATC
Phenotype
MIM: 603421
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MSANTD3-TMEFF1 PubMed Links

Gene Details

Gene
MSANTD3-TMEFF1
Gene Name
MSANTD3-TMEFF1 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198812.1 Intron NP_001185741.1
Gene
TMEFF1
Gene Name
transmembrane protein with EGF like and two follistatin like domains 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003692.4 Intron NP_003683.2

View Full Product Details