Product Details

SNP ID
rs451548
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:17791326 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGCCGAGAGAACGCTTGTGTCGG[C/G]TGCAGGGAGTGCCGCGCCCACCCTG
Phenotype
MIM: 608882
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MICAL3 PubMed Links

Gene Details

Gene
MICAL3
Gene Name
microtubule associated monooxygenase, calponin and LIM domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001122731.2 Intron NP_001116203.1
NM_001136004.3 Intron NP_001129476.1
NM_015241.2 Intron NP_056056.2
XM_005261262.3 Intron XP_005261319.1
XM_011546137.2 Intron XP_011544439.1
XM_011546139.2 Intron XP_011544441.1
XM_011546142.2 Intron XP_011544444.1
XM_011546146.2 Intron XP_011544448.1
XM_017028865.1 Intron XP_016884354.1
XM_017028866.1 Intron XP_016884355.1
XM_017028867.1 Intron XP_016884356.1
XM_017028868.1 Intron XP_016884357.1
XM_017028869.1 Intron XP_016884358.1
XM_017028870.1 Intron XP_016884359.1
XM_017028871.1 Intron XP_016884360.1
XM_017028872.1 Intron XP_016884361.1
XM_017028873.1 Intron XP_016884362.1
XM_017028874.1 Intron XP_016884363.1
XM_017028875.1 Intron XP_016884364.1
XM_017028876.1 Intron XP_016884365.1
XM_017028877.1 Intron XP_016884366.1
XM_017028878.1 Intron XP_016884367.1
XM_017028879.1 Intron XP_016884368.1
XM_017028880.1 Intron XP_016884369.1
XM_017028881.1 Intron XP_016884370.1
XM_017028882.1 Intron XP_016884371.1
XM_017028883.1 Intron XP_016884372.1
XM_017028884.1 Intron XP_016884373.1
XM_017028885.1 Intron XP_016884374.1
XM_017028886.1 Intron XP_016884375.1
XM_017028887.1 Intron XP_016884376.1

View Full Product Details