Product Details

SNP ID
rs71380229
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:984503 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGCCTCGGGTCTTCTCATAGCAC[A/C]GCGCTTCATGGAATTTTCTCGGCCG
Phenotype
MIM: 611761 MIM: 605923
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LMF1 PubMed Links

Gene Details

Gene
LMF1
Gene Name
lipase maturation factor 1
There are no transcripts associated with this gene.

Gene
SOX8
Gene Name
SRY-box 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014587.4 Intron NP_055402.2

View Full Product Details