Product Details

SNP ID
rs2238393
Assay Type
Functionally Tested
NCBI dbSNP Submissions
33
Location
Chr.16:3191975 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
CACGGCTGCCACAAAGAAAAGTAAA[G/T]GGCCTGCTGCTGTGGCTCGTTGGTC
Phenotype
MIM: 603232
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
OR1F1 PubMed Links

Gene Details

Gene
OR1F1
Gene Name
olfactory receptor family 1 subfamily F member 1
There are no transcripts associated with this gene.

View Full Product Details