Product Details

SNP ID
rs11690351
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:159239876 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGCCCTGAGAGACTGAGGTTCTGA[A/G]AAACTAAACTTCTGTTTCCTCCGTC
Phenotype
MIM: 611397
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TANC1 PubMed Links

Gene Details

Gene
TANC1
Gene Name
tetratricopeptide repeat, ankyrin repeat and coiled-coil containing 1
There are no transcripts associated with this gene.

Gene
WDSUB1
Gene Name
WD repeat, sterile alpha motif and U-box domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128212.2 Intron NP_001121684.1
NM_001128213.1 Intron NP_001121685.1
NM_001307994.1 Intron NP_001294923.1
NM_152528.2 Intron NP_689741.2
XM_005246355.2 Intron XP_005246412.2
XM_006712341.3 Intron XP_006712404.1
XM_011510753.2 Intron XP_011509055.1
XM_011510754.2 Intron XP_011509056.1
XM_017003481.1 Intron XP_016858970.1
XM_017003482.1 Intron XP_016858971.1
XM_017003483.1 Intron XP_016858972.1

View Full Product Details