Product Details

SNP ID
rs35505941
Assay Type
Functionally tested
NCBI dbSNP Submissions
21
Location
Chr.1:75786498 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCAGGCCTTCTTCTGTAGGGAGG[A/G]CCTGTTAAGAGTTGAGCAGCCCGAT
Phenotype
MIM: 179080
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RABGGTB PubMed Links

Gene Details

Gene
RABGGTB
Gene Name
Rab geranylgeranyltransferase beta subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004582.3 Intron NP_004573.2
Gene
SNORD45A
Gene Name
small nucleolar RNA, C/D box 45A
There are no transcripts associated with this gene.

Gene
SNORD45B
Gene Name
small nucleolar RNA, C/D box 45B
There are no transcripts associated with this gene.

Gene
SNORD45C
Gene Name
small nucleolar RNA, C/D box 45C
There are no transcripts associated with this gene.

View Full Product Details