Product Details

SNP ID
rs9694286
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:142655766 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTAAGCGCGTTCTGCCTCCTGTC[G/T]TCTTGGGTCCTCTCTTCCTCCTTTG
Phenotype
MIM: 603210
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
JRK PubMed Links

Gene Details

Gene
JRK
Gene Name
Jrk helix-turn-helix protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077527.2 11920 Intron NP_001070995.2
NM_001279352.1 11920 Intron NP_001266281.1
NM_003724.3 11920 Intron NP_003715.3
XM_006716677.3 11920 UTR 3 XP_006716740.1
XM_006716678.3 11920 UTR 3 XP_006716741.1
XM_011517354.1 11920 Intron XP_011515656.1
XM_011517355.1 11920 Intron XP_011515657.1

View Full Product Details