Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001033604.1 | Intron | NP_001028776.1 | |||
NM_001033605.1 | Intron | NP_001028777.1 | |||
NM_014451.3 | Intron | NP_055266.2 | |||
NM_198428.2 | Intron | NP_940820.1 | |||
XM_005249700.3 | Intron | XP_005249757.1 | |||
XM_005249701.2 | Intron | XP_005249758.1 | |||
XM_011515264.2 | Intron | XP_011513566.1 | |||
XM_011515265.2 | Intron | XP_011513567.1 | |||
XM_011515266.2 | Intron | XP_011513568.1 | |||
XM_011515267.2 | Intron | XP_011513569.1 | |||
XM_011515268.1 | Intron | XP_011513570.1 | |||
XM_011515269.2 | Intron | XP_011513571.1 | |||
XM_011515270.2 | Intron | XP_011513572.1 | |||
XM_017011990.1 | Intron | XP_016867479.1 | |||
XM_017011991.1 | Intron | XP_016867480.1 | |||
XM_017011992.1 | Intron | XP_016867481.1 | |||
XM_017011993.1 | Intron | XP_016867482.1 | |||
XM_017011994.1 | Intron | XP_016867483.1 |