Product Details

SNP ID
rs12552611
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:104765549 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATTTACCCATTGGGCATATGACTA[C/T]TACAAAGGGTGGTTCTTAAAGGATA
Phenotype
MIM: 608871 MIM: 608872
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NIPSNAP3A PubMed Links

Gene Details

Gene
NIPSNAP3A
Gene Name
nipsnap homolog 3A
There are no transcripts associated with this gene.

Gene
NIPSNAP3B
Gene Name
nipsnap homolog 3B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018376.3 Intron NP_060846.2
XM_011518839.1 Intron XP_011517141.1
XM_017014889.1 Intron XP_016870378.1

View Full Product Details