Product Details
- SNP ID
-
rs35163979
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:83657218 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAAATGGGGCTGAGGCTTCAAGGCA[T/C]AGTGTTCATTTTGCCTTCTTTCACC
- Phenotype
-
MIM: 609199
- Polymorphism
- T/C, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ADAMTSL3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs34875815] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ADAMTSL3
- Gene Name
- ADAMTS like 3
View Full Product Details