Product Details

SNP ID
rs1366333
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:123347318 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTGCTCCTAAACCTTTGTTGTTGT[A/T]TTGAACATATTTTTAAAAATTCATT
Phenotype
MIM: 613446
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CEP120 PubMed Links

Gene Details

Gene
CEP120
Gene Name
centrosomal protein 120
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166226.1 Intron NP_001159698.1
NM_153223.3 Intron NP_694955.2
XM_005271901.4 Intron XP_005271958.1
XM_011543185.2 Intron XP_011541487.1
XM_011543186.2 Intron XP_011541488.1
XM_017009085.1 Intron XP_016864574.1

View Full Product Details