Product Details

SNP ID
rs1356071
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:109136927 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCTATTTCTTTTTTATTTATCAT[A/G]TCTCCTACAGCCATCAGGTTGATAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FLJ22763 PubMed Links

Gene Details

Gene
FLJ22763
Gene Name
uncharacterized LOC401081
There are no transcripts associated with this gene.

View Full Product Details