Product Details

SNP ID
rs1207579
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:24363316 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGAAGATGATGACAAGAGATGGC[C/G]AGAAATGGAGCCAGGAAGACAAGCT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SUPT20HL1 PubMed Links

Gene Details

Gene
SUPT20HL1
Gene Name
SPT20 homolog, SAGA complex component-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136234.1 556 Missense Mutation CAG,GAG Q186E NP_001129706.1

View Full Product Details