Product Details

SNP ID
rs1043009
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:3777522 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACTTCTGAACGGCCGAAGGTGCC[C/T]CATTCCAGACCTGCCCATTTGATGG
Phenotype
MIM: 602053
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLF6 PubMed Links

Gene Details

Gene
KLF6
Gene Name
Kruppel like factor 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160124.1 3012 UTR 3 NP_001153596.1
NM_001160125.1 3012 UTR 3 NP_001153597.1
NM_001300.5 3012 UTR 3 NP_001291.3

View Full Product Details