Product Details

SNP ID
rs1319172
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:51601539 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTTAAGGTAGACTTCTGGCATTA[A/G]AAATTGCTCAAAAGCTTTATAGGTC
Phenotype
MIM: 600702
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SCN8A PubMed Links

Gene Details

Gene
SCN8A
Gene Name
sodium voltage-gated channel alpha subunit 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001177984.2 Intron NP_001171455.1
NM_014191.3 Intron NP_055006.1
XM_006719556.3 Intron XP_006719619.1
XM_011538650.2 Intron XP_011536952.1
XM_011538651.2 Intron XP_011536953.1
XM_017019794.1 Intron XP_016875283.1
XM_017019795.1 Intron XP_016875284.1
XM_017019796.1 Intron XP_016875285.1

View Full Product Details