Product Details

SNP ID
rs1608100
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:158115844 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCCTTTTCAGGTAATTATGTCTTT[C/T]GTATTACACCAAAAATTGATTGTGT
Phenotype
MIM: 613352 MIM: 602504
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RSRC1 PubMed Links

Gene Details

Gene
RSRC1
Gene Name
arginine and serine rich coiled-coil 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271834.1 Intron NP_001258763.1
NM_001271838.1 Intron NP_001258767.1
NM_016625.3 Intron NP_057709.2
Gene
SHOX2
Gene Name
short stature homeobox 2
There are no transcripts associated with this gene.

View Full Product Details