Product Details
- SNP ID
-
rs786428
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:123602451 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGCCAGCGAGCCGGCAGAGGGGCCG[C/G]GACCCACCCCGCCCTCGGGTGCTGT
- Phenotype
-
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
DDX55
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78690737] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DDX55
- Gene Name
- DEAD-box helicase 55
- Gene
- TMED2
- Gene Name
- transmembrane p24 trafficking protein 2
There are no transcripts associated with this gene.
View Full Product Details