Product Details

SNP ID
rs622463
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:28648556 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTCAAAGGATTAAGCAGCTAATGA[A/C]ATAGCACACCTAATAAATAACAACA
Phenotype
MIM: 610664 MIM: 602934
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KIAA0100 PubMed Links

Gene Details

Gene
KIAA0100
Gene Name
KIAA0100
There are no transcripts associated with this gene.

Gene
SDF2
Gene Name
stromal cell derived factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006923.3 1365 UTR 3 NP_008854.2
XM_011525106.2 1365 Intron XP_011523408.1

View Full Product Details