Product Details

SNP ID
rs1048167
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.5:74721674 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AAACATTTAGGTCAAACCACTTCTC[C/T]GGTTGAGCAGTGTATTTTGATCTTG
Phenotype
MIM: 606544 MIM: 606873
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GFM2 PubMed Links

Gene Details

Gene
GFM2
Gene Name
G elongation factor mitochondrial 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281302.1 2673 Missense Mutation NP_001268231.1
NM_032380.4 2673 Missense Mutation NP_115756.2
NM_170681.2 2673 Intron NP_733781.1
NM_170691.2 2673 Missense Mutation NP_733792.1
XM_011543691.2 2673 Intron XP_011541993.1
XM_017009986.1 2673 Missense Mutation XP_016865475.1
Gene
HEXB
Gene Name
hexosaminidase subunit beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000521.3 2673 Intron NP_000512.1
NM_001292004.1 2673 Intron NP_001278933.1

View Full Product Details