Product Details

SNP ID
rs10459
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:76311656 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TATCCACAGTCTTTATCTTGGGACA[C/T]TGCAGCTTCTGAACCTCATGAGGGA
Phenotype
MIM: 601249
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRPSAP1 PubMed Links

Gene Details

Gene
PRPSAP1
Gene Name
phosphoribosyl pyrophosphate synthetase associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002766.2 1490 Silent Mutation CAA,CAG Q348Q NP_002757.2
XM_011525034.2 1490 Silent Mutation CAA,CAG Q245Q XP_011523336.1
Gene
QRICH2
Gene Name
glutamine rich 2
There are no transcripts associated with this gene.

View Full Product Details