Product Details

SNP ID
rs708563
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:45263569 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATGCTCCTACAGTGTCACTGCCCT[A/G]CCCTGCCCTGCCAGAGGGCAGCCAT
Phenotype
MIM: 604655
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
MAP3K14 PubMed Links

Gene Details

Gene
MAP3K14
Gene Name
mitogen-activated protein kinase kinase kinase 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003954.4 4173 UTR 3 NP_003945.2
XM_011525441.2 4173 UTR 3 XP_011523743.1
Gene
MAP3K14-AS1
Gene Name
MAP3K14 antisense RNA 1
There are no transcripts associated with this gene.

Gene
SPATA32
Gene Name
spermatogenesis associated 32
There are no transcripts associated with this gene.

View Full Product Details