Product Details

SNP ID
rs1420111
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:29564906 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCCGAGGCCGCCCCGCCGCCTCC[C/G]GTAGGCGATCAGACTTCCTCTCCCT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PRR15 PubMed Links

Gene Details

Gene
PRR15
Gene Name
proline rich 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175887.2 523 Intron NP_787083.1
XM_005249653.3 523 UTR 5 XP_005249710.1
XM_011515198.1 523 UTR 5 XP_011513500.1
XM_011515199.1 523 Intron XP_011513501.1
XM_017011842.1 523 Intron XP_016867331.1

View Full Product Details