Product Details

SNP ID
rs1044332
Assay Type
Validated
NCBI dbSNP Submissions
34
Location
Chr.1:20499750 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
TGAGTACACCAAGATGTCAAAGCTG[C/T]GACGGGCTGGATGAGGGAGCCCCAA
Phenotype
MIM: 612037
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MUL1 PubMed Links

Gene Details

Gene
MUL1
Gene Name
mitochondrial E3 ubiquitin protein ligase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024544.2 2839 UTR 3 NP_078820.2
XM_011542137.2 2839 UTR 3 XP_011540439.1

View Full Product Details