Product Details
- SNP ID
-
rs3096
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:47952066 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- AGACATGAGCATAAAAGAGGTTCTC[G/T]GCCTTTCCAGCGTTGTTATTACAGA
- Phenotype
-
MIM: 603287
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
PNPO
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115159645] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PNPO
- Gene Name
- pyridoxamine 5'-phosphate oxidase
There are no transcripts associated with this gene.
- Gene
- PRR15L
- Gene Name
- proline rich 15 like
View Full Product Details