Product Details

SNP ID
rs6905
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:73283913 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCATGGGGACGCCCCTGGCTGTGGT[A/T]GGTTCTGTGTCTCCCCCTCCCCTCA
Phenotype
MIM: 605468
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CDC42EP4 PubMed Links

Gene Details

Gene
CDC42EP4
Gene Name
CDC42 effector protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012121.4 3127 UTR 3 NP_036253.2
XM_005257182.2 3127 UTR 3 XP_005257239.1

View Full Product Details