Product Details

SNP ID
rs3476
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:18224942 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAAATTACAAATTGATGACACTTT[C/T]GAGGCAAAGCAGGGTAACTGTTCCT
Phenotype
MIM: 125264 MIM: 613081
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DEK PubMed Links

Gene Details

Gene
DEK
Gene Name
DEK proto-oncogene
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134709.1 1996 UTR 3 NP_001128181.1
NM_003472.3 1996 UTR 3 NP_003463.1
XM_017011273.1 1996 Intron XP_016866762.1
Gene
KDM1B
Gene Name
lysine demethylase 1B
There are no transcripts associated with this gene.

View Full Product Details