Product Details

SNP ID
rs913648
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:92452985 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAGCCCCATAGTTGTGACTATATC[A/G]TTCTTGCCCAAAATTGAGATTTGGA
Phenotype
MIM: 146680
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IDE PubMed Links

Gene Details

Gene
IDE
Gene Name
insulin degrading enzyme
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001165946.1 3179 UTR 3 NP_001159418.1
NM_001322793.1 3179 UTR 3 NP_001309722.1
NM_001322794.1 3179 UTR 3 NP_001309723.1
NM_001322795.1 3179 UTR 3 NP_001309724.1
NM_001322796.1 3179 UTR 3 NP_001309725.1
NM_001322797.1 3179 UTR 3 NP_001309726.1
NM_004969.3 3179 UTR 3 NP_004960.2
XM_017016187.1 3179 UTR 3 XP_016871676.1
XM_017016188.1 3179 UTR 3 XP_016871677.1
XM_017016189.1 3179 UTR 3 XP_016871678.1
XM_017016190.1 3179 UTR 3 XP_016871679.1

View Full Product Details