Product Details

SNP ID
rs836178
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:50104356 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTTTGGGGGTGGGAAGATGAGAAG[G/T]GGGGCTCCCTTCTCTAGTAGCAAAG
Phenotype
MIM: 614478 MIM: 138420 MIM: 601735
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
COX14 PubMed Links

Gene Details

Gene
COX14
Gene Name
COX14, cytochrome c oxidase assembly factor
There are no transcripts associated with this gene.

Gene
GPD1
Gene Name
glycerol-3-phosphate dehydrogenase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001257199.1 Intron NP_001244128.1
NM_005276.3 Intron NP_005267.2
Gene
SMARCD1
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 1
There are no transcripts associated with this gene.

View Full Product Details