Product Details

SNP ID
rs1803474
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:29073803 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACACAGAGTGAGGACTCATGTCTA[A/T]TGAGCACCGGCCAAAACTTCCATCT
Phenotype
MIM: 610067 MIM: 609517
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MYO18A PubMed Links

Gene Details

Gene
MYO18A
Gene Name
myosin XVIIIA
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_078471.3 1807 UTR 3 NP_510880.2
NM_203318.1 1807 UTR 3 NP_976063.1
Gene
TIAF1
Gene Name
TGFB1-induced anti-apoptotic factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004740.3 1807 UTR 3 NP_004731.2

View Full Product Details