Product Details

SNP ID
rs899323
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:75173799 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GGCTCTCATGAGCCACCCTGCTAGG[A/C]CAAAACTGGAAGTATTTAATGGAAC
Phenotype
MIM: 603042
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SUMO2 PubMed Links

Gene Details

Gene
SUMO2
Gene Name
small ubiquitin-like modifier 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005849.1 Intron NP_001005849.1
NM_006937.3 Intron NP_008868.3

View Full Product Details