Product Details
- SNP ID
-
rs1517351
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:191031319 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAGTGTGCCCTGAAGGCTAGCCTTA[G/T]GTATTAAAGGAAATGGGACATTGCA
- Phenotype
-
MIM: 600558
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC105373805
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3024900] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC105373805
- Gene Name
- uncharacterized LOC105373805
There are no transcripts associated with this gene.
- Gene
- STAT4
- Gene Name
- signal transducer and activator of transcription 4
View Full Product Details