Product Details

SNP ID
rs1135326
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:159872559 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCAGTACTTCTCGGGAAGGCCAG[G/T]GGCTCTGCCGGGGAAACGCAGGCAG
Phenotype
MIM: 605152
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CFAP45 PubMed Links

Gene Details

Gene
CFAP45
Gene Name
cilia and flagella associated protein 45
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012337.2 1600 Missense Mutation ACT,CCT T528P NP_036469.2
Gene
VSIG8
Gene Name
V-set and immunoglobulin domain containing 8
There are no transcripts associated with this gene.

View Full Product Details