Product Details
- SNP ID
-
rs1046196
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:189746782 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CAGATTTTCCTTAGCATGTCTACAG[A/G]AATTTAGTGTCAGGTCAGAGTTATG
- Phenotype
-
MIM: 609803
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ANKAR
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs201037870,rs80306810] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ANKAR
- Gene Name
- ankyrin and armadillo repeat containing
- Gene
- OSGEPL1
- Gene Name
- O-sialoglycoprotein endopeptidase-like 1
View Full Product Details