Product Details

SNP ID
rs1130295
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:104769787 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCCCCATCCCTGGTCCCATCCCA[A/G]GGGCCCAGCCTCCGATGACGTCCTC
Phenotype
MIM: 164730
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AKT1 PubMed Links

Gene Details

Gene
AKT1
Gene Name
AKT serine/threonine kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014431.1 2337 UTR 3 NP_001014431.1
NM_001014432.1 2337 UTR 3 NP_001014432.1
NM_005163.2 2337 UTR 3 NP_005154.2
XM_005267401.1 2337 UTR 3 XP_005267458.1
XM_017021075.1 2337 Intron XP_016876564.1
XM_017021076.1 2337 Intron XP_016876565.1
XM_017021077.1 2337 Intron XP_016876566.1
XM_017021078.1 2337 Intron XP_016876567.1

View Full Product Details