Product Details

SNP ID
rs4831
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:186613180 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCTGCTCCTTTGTCTTGCTCAGCT[C/G]TGGGGCTGCCACTCAGCCCCACATG
Phenotype
MIM: 138680
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
AHSG PubMed Links

Gene Details

Gene
AHSG
Gene Name
alpha 2-HS glycoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001622.2 253 Silent Mutation CTC,CTG L13L NP_001613.2
XM_017005840.1 253 Silent Mutation CTC,CTG L13L XP_016861329.1

View Full Product Details