Product Details

SNP ID
rs1135867
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:90964283 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTACGAATATAAGTCAAATTTTTA[A/C]CATTGCAGAACGTATCTTCGTGCAT
Phenotype
MIM: 601657
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CCER1 PubMed Links

Gene Details

Gene
CCER1
Gene Name
coiled-coil glutamate rich protein 1
There are no transcripts associated with this gene.

Gene
EPYC
Gene Name
epiphycan
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004950.4 751 Missense Mutation GGT,GTT G281V NP_004941.2
XM_011538008.1 751 Missense Mutation GGT,GTT G220V XP_011536310.1

View Full Product Details