Product Details
- SNP ID
-
rs1056948
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:53567166 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATAAAACAAGTAATGTCTACTGTT[C/T]GACAACTTAAGTATCAACAATTAAA
- Phenotype
-
MIM: 602967
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC101927770
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75839044] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC101927770
- Gene Name
- uncharacterized LOC101927770
There are no transcripts associated with this gene.
- Gene
- ZNF217
- Gene Name
- zinc finger protein 217
View Full Product Details