Product Details

SNP ID
rs1555926
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:53568405 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGTCCAACAAAAATGATTAATTTGA[C/T]AGAAACAAATTTAAATAACTGCACA
Phenotype
MIM: 602967
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101927770 PubMed Links

Gene Details

Gene
LOC101927770
Gene Name
uncharacterized LOC101927770
There are no transcripts associated with this gene.

Gene
ZNF217
Gene Name
zinc finger protein 217
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006526.2 4398 UTR 3 NP_006517.1
XM_005260545.3 4398 UTR 3 XP_005260602.1
XM_006723875.2 4398 UTR 3 XP_006723938.1
XM_011529036.1 4398 UTR 3 XP_011527338.1
XM_017028059.1 4398 UTR 3 XP_016883548.1

View Full Product Details