Product Details

SNP ID
rs1076063
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:149732632 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGGCCGAGGATCTTCTGGTGATCT[A/T]TGCCGGCCCAACTTGGGAAATGTAA
Phenotype
MIM: 611957 MIM: 608886
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MIR378A PubMed Links

Gene Details

Gene
MIR378A
Gene Name
microRNA 378a
There are no transcripts associated with this gene.

Gene
PPARGC1B
Gene Name
PPARG coactivator 1 beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172698.1 Intron NP_001166169.1
NM_001172699.1 Intron NP_001166170.1
NM_133263.3 Intron NP_573570.3
XM_005268372.4 Intron XP_005268429.1
XM_011537553.2 Intron XP_011535855.1
XM_011537554.2 Intron XP_011535856.1
XM_011537555.2 Intron XP_011535857.1
XM_011537556.2 Intron XP_011535858.1
XM_011537557.1 Intron XP_011535859.1

View Full Product Details