Product Details

SNP ID
rs13078
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.14:95090410 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TGCCTTCAATTCATTCCACTCACTA[A/T]CAACTTTAAGTCTTCCTTTCCGATT
Phenotype
MIM: 606241
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DICER1 PubMed Links

Gene Details

Gene
DICER1
Gene Name
dicer 1, ribonuclease III
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195573.1 5978 UTR 3 NP_001182502.1
NM_001271282.2 5978 UTR 3 NP_001258211.1
NM_001291628.1 5978 UTR 3 NP_001278557.1
NM_030621.4 5978 UTR 3 NP_085124.2
NM_177438.2 5978 UTR 3 NP_803187.1
XM_011536599.1 5978 UTR 3 XP_011534901.1
XM_011536600.2 5978 UTR 3 XP_011534902.1
XM_011536601.2 5978 UTR 3 XP_011534903.1
XM_011536602.2 5978 UTR 3 XP_011534904.1
XM_011536604.2 5978 UTR 3 XP_011534906.1
XM_011536605.2 5978 UTR 3 XP_011534907.1
XM_017021120.1 5978 UTR 3 XP_016876609.1
XM_017021121.1 5978 UTR 3 XP_016876610.1
XM_017021122.1 5978 UTR 3 XP_016876611.1
XM_017021123.1 5978 UTR 3 XP_016876612.1

View Full Product Details