Product Details

SNP ID
rs1037448
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.11:61367060 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GCAGGTGGATGTTGATACAGGTTAT[C/T]CTTAGGCATCAGTGCCCCATGGCTA
Phenotype
MIM: 614459
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CYB561A3 PubMed Links

Gene Details

Gene
CYB561A3
Gene Name
cytochrome b561 family member A3
There are no transcripts associated with this gene.

Gene
TMEM138
Gene Name
transmembrane protein 138
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016464.4 Intron NP_057548.1
XM_006718585.3 Intron XP_006718648.1
XM_006718586.2 Intron XP_006718649.1
XM_006718588.3 Intron XP_006718651.1
XM_011545098.2 Intron XP_011543400.1
XM_011545099.1 Intron XP_011543401.1
XM_011545100.1 Intron XP_011543402.1
XM_017017917.1 Intron XP_016873406.1

View Full Product Details