Product Details

SNP ID
rs982714
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:152391542 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATCACTGACCACAGATCACCATAAG[C/T]ACATATAATAATAATCAAGTTTATT
Phenotype
MIM: 605108
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
NMUR2 PubMed Links

Gene Details

Gene
NMUR2
Gene Name
neuromedin U receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020167.4 2063 UTR 3 NP_064552.3
XM_011537670.2 2063 Intron XP_011535972.1

View Full Product Details