Product Details

SNP ID
rs1534176
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:36954747 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGACCCCTGGACGGGAGAAGGAGA[A/G]ACGGCTGGCCACCATGCACGGCTCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PI16 PubMed Links

Gene Details

Gene
PI16
Gene Name
peptidase inhibitor 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199159.1 210 UTR 5 NP_001186088.1
NM_153370.2 210 UTR 5 NP_699201.2
XM_005248917.2 210 UTR 5 XP_005248974.1
XM_011514375.2 210 UTR 5 XP_011512677.1
XM_017010430.1 210 UTR 5 XP_016865919.1

View Full Product Details