Product Details

SNP ID
rs1128881
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:150693290 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTCCATCTTTCTCTTTTTCTTTT[C/T]CTCTGGCTGCCTGGGATCTGGGAAA
Phenotype
MIM: 612430
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RBM22 PubMed Links

Gene Details

Gene
RBM22
Gene Name
RNA binding motif protein 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018047.2 1051 Missense Mutation GAA,GGA E310G NP_060517.1

View Full Product Details