Product Details

SNP ID
rs1062309
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31662218 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGTCAAAGGGCCCAGGTCAGAG[A/G]CCCAAGTTCAGACTTCAGCAGCAGA
Phenotype
MIM: 606907 MIM: 142590 MIM: 115441 MIM: 142610 MIM: 610433
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
APOM PubMed Links

Gene Details

Gene
APOM
Gene Name
apolipoprotein M
There are no transcripts associated with this gene.

Gene
BAG6
Gene Name
BCL2 associated athanogene 6
There are no transcripts associated with this gene.

Gene
C6orf47
Gene Name
chromosome 6 open reading frame 47
There are no transcripts associated with this gene.

Gene
CSNK2B
Gene Name
casein kinase 2 beta
There are no transcripts associated with this gene.

Gene
GPANK1
Gene Name
G-patch domain and ankyrin repeats 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199237.1 1780 UTR 3 NP_001186166.1
NM_001199238.1 1780 UTR 3 NP_001186167.1
NM_001199239.1 1780 UTR 3 NP_001186168.1
NM_001199240.1 1780 UTR 3 NP_001186169.1
NM_033177.3 1780 UTR 3 NP_149417.1
XM_005249403.3 1780 UTR 3 XP_005249460.1
XM_006715204.1 1780 UTR 3 XP_006715267.1
XM_011514909.1 1780 UTR 3 XP_011513211.1
XM_011514910.1 1780 UTR 3 XP_011513212.1
Gene
LY6G5B
Gene Name
lymphocyte antigen 6 complex, locus G5B
There are no transcripts associated with this gene.

View Full Product Details